Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ca14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328144.1 | NP_001315073.1 | 1 - 2 | 221 | 133 | |
NR_137291.1 | 1 - 2 | 221 | 133 | |||
GenBank mRNA | BC107639.1 | - | 1 - 2 | 214 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |