Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: foxp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039637.1 | NP_001034726.1 | 18 - 19 | 1958 | 78 | |
XM_005166055.4 | XP_005166112.1 | 19 - 20 | 2392 | 78 | ||
XM_021476910.1 | XP_021332585.1 | 19 - 20 | 2091 | 78 | ||
XM_021476911.1 | XP_021332586.1 | 18 - 19 | 2352 | 78 | ||
XM_021476912.1 | XP_021332587.1 | 16 - 17 | 2184 | 78 | ||
XM_021476913.1 | XP_021332588.1 | 15 - 16 | 1747 | 78 | ||
XM_021476914.1 | XP_021332589.1 | 15 - 16 | 1844 | 78 | ||
XM_021476916.1 | XP_021332591.1 | 15 - 16 | 1981 | 78 | ||
XM_021476917.1 | XP_021332592.1 | 19 - 20 | 2390 | 78 | ||
XM_021476918.1 | XP_021332593.1 | 14 - 15 | 1672 | 78 | ||
GenBank mRNA | BC163445.1 | - | 15 - 16 | 1709 | 78 | |
DQ333303.2 | - | 18 - 19 | 1958 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |