Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: foxp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039637.1 | NP_001034726.1 | 19 - 20 | 2120 | 82 | |
XM_005166055.4 | XP_005166112.1 | 20 - 21 | 2554 | 82 | ||
XM_021476910.1 | XP_021332585.1 | 20 - 21 | 2253 | 82 | ||
XM_021476911.1 | XP_021332586.1 | 19 - 20 | 2514 | 82 | ||
XM_021476912.1 | XP_021332587.1 | 17 - 18 | 2346 | 82 | ||
XM_021476913.1 | XP_021332588.1 | 16 - 17 | 1909 | 82 | ||
XM_021476914.1 | XP_021332589.1 | 16 - 17 | 2006 | 82 | ||
XM_021476916.1 | XP_021332591.1 | 16 - 17 | 2143 | 82 | ||
XM_021476917.1 | XP_021332592.1 | 20 - 21 | 2552 | 82 | ||
XM_021476918.1 | XP_021332593.1 | 15 - 16 | 1834 | 82 | ||
GenBank mRNA | BC163445.1 | - | 16 - 17 | 1871 | 82 | |
DQ333303.2 | - | 19 - 20 | 2120 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |