Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scn4aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039825.1 | NP_001034914.1 | 13 - 14 | 2034 | 64 | |
XM_017358479.2 | XP_017213968.1 | 14 - 15 | 2220 | 64 | ||
GenBank mRNA | DQ149506.1 | - | 13 - 14 | 2034 | 64 | |
DQ351532.1 | - | 2 - 3 | 222 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Danio rerio Probe spans exons |