Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scn4aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039825.1 | NP_001034914.1 | 14 - 15 | 2386 | 84 | |
XM_017358479.2 | XP_017213968.1 | 15 - 16 | 2572 | 84 | ||
GenBank mRNA | DQ149506.1 | - | 14 - 15 | 2386 | 84 | |
DQ351532.1 | - | 3 - 4 | 574 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |