Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cnpy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039826.2 | NP_001034915.2 | 2 - 3 | 150 | 66 | |
GenBank mRNA | AB201384.1 | - | 2 - 3 | 125 | 66 | |
BC121730.1 | - | 2 - 3 | 150 | 66 | ||
BC163941.1 | - | 1 - 2 | 133 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |