Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gsg1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039976.1 | NP_001035065.1 | 2 - 3 | 564 | 62 | |
GenBank mRNA | BC097105.1 | - | 2 - 3 | 564 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |