Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: edn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045185.1 | NP_001038650.1 | 3 - 4 | 407 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |