Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: itln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076622.2 | NP_001070090.1 | 4 - 5 | 451 | 147 | |
XM_017355768.2 | XP_017211257.1 | 4 - 5 | 420 | 147 | ||
GenBank mRNA | BC124294.1 | - | 4 - 5 | 454 | 147 | |
BC164264.1 | - | 3 - 4 | 440 | 147 | ||
EU583680.1 | - | 3 - 4 | 434 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |