Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bmp7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077146.2 | NP_001070614.2 | 3 - 4 | 926 | 117 | |
GenBank mRNA | BC124215.1 | - | 3 - 4 | 932 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |