Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dcaf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077580.1 | NP_001071048.1 | 6 - 7 | 1152 | 81 | |
GenBank mRNA | BC124796.1 | - | 6 - 7 | 1152 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |