Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nop53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079946.1 | NP_001073415.1 | 1 - 2 | 237 | 71 | |
GenBank mRNA | BC109456.1 | - | 1 - 2 | 237 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |