Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nrxn2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079984.1 | NP_001073453.1 | 13 - 14 | 4824 | 97 | |
GenBank mRNA | DQ641430.1 | - | 13 - 14 | 4824 | 97 | |
DQ641431.1 | - | 6 - 7 | 2099 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Danio rerio Probe spans exons |