Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nrxn1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080021.2 | NP_001073490.1 | 16 - 17 | 4192 | 88 | |
XM_009306651.3 | XP_009304926.1 | 16 - 17 | 4173 | 88 | ||
XM_009306654.3 | XP_009304929.1 | 16 - 17 | 4174 | 88 | ||
XM_009306656.3 | XP_009304931.1 | 16 - 17 | 4174 | 88 | ||
XM_021480137.1 | XP_021335812.1 | 16 - 17 | 4173 | 88 | ||
XM_021480139.1 | XP_021335814.1 | 15 - 16 | 4147 | 88 | ||
XM_021480140.1 | XP_021335815.1 | 14 - 15 | 4137 | 88 | ||
XM_021480141.1 | XP_021335816.1 | 15 - 16 | 4147 | 88 | ||
XM_021480142.1 | XP_021335817.1 | 11 - 12 | 4063 | 88 | ||
XM_021480143.1 | XP_021335818.1 | 14 - 15 | 4143 | 88 | ||
XM_021480144.1 | XP_021335819.1 | 12 - 13 | 4086 | 88 | ||
XM_021480145.1 | XP_021335820.1 | 11 - 12 | 4063 | 88 | ||
XM_021480146.1 | XP_021335821.1 | 12 - 13 | 4087 | 88 | ||
GenBank mRNA | DQ641424.1 | - | 15 - 16 | 4141 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |