Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sema4e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083865.1 | NP_001077334.1 | 6 - 7 | 636 | 75 | |
NM_131049.1 | NP_571124.1 | 8 - 9 | 1180 | 75 | ||
XM_005168429.4 | XP_005168486.1 | 8 - 9 | 1169 | 75 | ||
XM_017359054.2 | XP_017214543.1 | 8 - 9 | 1148 | 75 | ||
XM_021481386.1 | XP_021337061.1 | 7 - 8 | 1110 | 75 | ||
GenBank mRNA | AF073289.1 | - | 8 - 9 | 1180 | 75 | |
BC078280.1 | - | 7 - 8 | 785 | 75 | ||
BC155168.1 | - | 8 - 9 | 1143 | 75 | ||
CU458994.1 | - | 6 - 7 | 636 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |