Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: st8sia4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099416.1 | NP_001092886.1 | 2 - 3 | 697 | 84 | |
GenBank mRNA | AJ715545.1 | - | 2 - 3 | 241 | 84 | |
BC098613.1 | - | 2 - 3 | 230 | 84 | ||
DQ779599.1 | - | 2 - 3 | 697 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |