Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: st8sia4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099416.1 | NP_001092886.1 | 3 - 4 | 953 | 86 | |
GenBank mRNA | AJ705099.1 | - | 1 - 2 | 257 | 86 | |
AJ715545.1 | - | 3 - 4 | 497 | 86 | ||
DQ779599.1 | - | 3 - 4 | 953 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |