Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: antxr2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102568.1 | NP_001096038.1 | 4 - 5 | 367 | 70 | |
GenBank mRNA | EF591979.1 | - | 4 - 5 | 367 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Danio rerio Probe spans exons |