Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myog | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131006.1 | NP_571081.1 | 2 - 3 | 764 | 76 | |
GenBank mRNA | AF202639.1 | - | 2 - 3 | 764 | 76 | |
BC078421.1 | - | 2 - 3 | 721 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Probe spans exons |