Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kita | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131053.1 | NP_571128.1 | 18 - 19 | 2613 | 123 | |
XM_009294634.3 | XP_009292909.1 | 18 - 19 | 2658 | 123 | ||
XM_009294635.3 | XP_009292910.1 | 18 - 19 | 2688 | 123 | ||
GenBank mRNA | AF153446.1 | - | 18 - 19 | 2613 | 123 | |
BC162502.1 | - | 18 - 19 | 2628 | 123 | ||
CU458898.1 | - | 18 - 19 | 2563 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |