Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pax2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131184.2 | NP_571259.1 | 9 - 10 | 1601 | 63 | |
XM_005156747.4 | XP_005156804.1 | 10 - 11 | 2282 | 63 | ||
XM_017358685.2 | XP_017214174.1 | 10 - 11 | 2465 | 63 | ||
XM_021480495.1 | XP_021336170.1 | 12 - 13 | 3164 | 63 | ||
XM_021480501.1 | XP_021336176.1 | 10 - 11 | 2479 | 63 | ||
XM_021480503.1 | XP_021336178.1 | 11 - 12 | 2948 | 63 | ||
XM_021480507.1 | XP_021336182.1 | 10 - 11 | 2295 | 63 | ||
XM_021480509.1 | XP_021336184.1 | 9 - 10 | 2244 | 63 | ||
XM_021480512.1 | XP_021336187.1 | 7 - 8 | 1953 | 63 | ||
XM_021480513.1 | XP_021336188.1 | 7 - 8 | 1940 | 63 | ||
GenBank mRNA | AF067530.1 | - | 10 - 11 | 1142 | 63 | |
AF067531.1 | - | 9 - 10 | 1097 | 63 | ||
AF067533.1 | - | 8 - 9 | 977 | 63 | ||
AF067534.1 | - | 10 - 11 | 1154 | 63 | ||
AF067535.1 | - | 7 - 8 | 800 | 63 | ||
AF067541.1 | - | 9 - 10 | 1109 | 63 | ||
BC066690.1 | - | 9 - 10 | 1601 | 63 | ||
X63961.1 | - | 1723 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |