Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: irx1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131823.1 | NP_571898.1 | 2 - 3 | 1334 | 98 | |
GenBank mRNA | AF414134.1 | - | 3 - 4 | 1295 | 98 | |
AY017308.1 | - | 2 - 3 | 1334 | 98 | ||
BC077101.1 | - | 2 - 3 | 1336 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |