Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: irx1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131823.1 | NP_571898.1 | 3 - 4 | 1414 | 64 | |
GenBank mRNA | AF414134.1 | - | 4 - 5 | 1375 | 64 | |
AY017308.1 | - | 3 - 4 | 1414 | 64 | ||
BC077101.1 | - | 3 - 4 | 1416 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |