Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: klf2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131856.3 | NP_571931.2 | 2 - 3 | 1286 | 69 | |
GenBank mRNA | AF392992.1 | - | 2 - 3 | 69 | ||
BC063933.1 | - | 2 - 3 | 1239 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Inventoried Probe spans exons |