Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npffl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183346.3 | NP_899189.2 | 2 - 3 | 279 | 78 | |
GenBank mRNA | AB193140.1 | - | 2 - 3 | 279 | 78 | |
AY092774.1 | - | 1 - 2 | 119 | 78 | ||
BC094983.1 | - | 2 - 3 | 297 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |