Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fen1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328507.1 | NP_001315436.1 | 11 - 12 | 1195 | 104 | |
XM_017353804.1 | XP_017209293.1 | 11 - 12 | 1213 | 104 | ||
GenBank mRNA | AY391423.1 | - | 11 - 12 | 1124 | 104 | |
AY648844.1 | - | 11 - 12 | 1137 | 104 | ||
BC049413.1 | - | 11 - 12 | 1412 | 104 | ||
BC071488.1 | - | 11 - 12 | 1156 | 104 | ||
CT678576.2 | - | 1 - 2 | 248 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |