Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199558.2 | NP_955852.2 | 15 - 16 | 2675 | 76 | |
GenBank mRNA | BC059198.1 | - | 15 - 16 | 2652 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |