Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199558.2 | NP_955852.2 | 17 - 18 | 2875 | 66 | |
GenBank mRNA | BC059198.1 | - | 17 - 18 | 2852 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |