Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tdo2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199856.1 | NP_956150.1 | 11 - 12 | 1141 | 68 | |
XM_021479554.1 | XP_021335229.1 | 11 - 12 | 1145 | 68 | ||
GenBank mRNA | BC055123.1 | - | 11 - 12 | 1141 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |