Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ndrg1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200692.3 | NP_956986.2 | 7 - 8 | 655 | 110 | |
GenBank mRNA | BC059429.1 | - | 7 - 8 | 600 | 110 | |
LC102484.1 | - | 7 - 8 | 655 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |