Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fkbp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200812.2 | NP_957106.1 | 3 - 4 | 283 | 106 | |
GenBank mRNA | BC059689.1 | - | 3 - 4 | 273 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |