Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cox15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201206.2 | NP_957500.2 | 6 - 7 | 937 | 57 | |
GenBank mRNA | BC046017.1 | - | 6 - 7 | 924 | 57 | |
BC066452.1 | - | 6 - 7 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |