Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ncln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207057.1 | NP_996940.1 | 13 - 14 | 1686 | 91 | |
XM_005161589.4 | XP_005161646.1 | 13 - 14 | 91 | |||
XM_005161590.4 | XP_005161647.1 | 13 - 14 | 91 | |||
XM_005161591.4 | XP_005161648.1 | 13 - 14 | 91 | |||
GenBank mRNA | BC056523.1 | - | 13 - 14 | 1676 | 91 | |
BC066393.1 | - | 13 - 14 | 1686 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |