Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ilf3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212599.1 | NP_997764.1 | 4 - 5 | 514 | 98 | |
XM_005163739.4 | XP_005163796.1 | 4 - 5 | 586 | 98 | ||
XM_005163740.4 | XP_005163797.1 | 4 - 5 | 586 | 98 | ||
XM_009299225.3 | XP_009297500.1 | 4 - 5 | 586 | 98 | ||
GenBank mRNA | BC047175.1 | - | 4 - 5 | 543 | 98 | |
BC067172.1 | - | 4 - 5 | 514 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |