Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nrp2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212965.1 | NP_998130.1 | 3 - 4 | 1048 | 86 | |
XM_021469092.1 | XP_021324767.1 | 3 - 4 | 1068 | 86 | ||
GenBank mRNA | AY351912.1 | - | 3 - 4 | 784 | 86 | |
AY428801.1 | - | 3 - 4 | 1048 | 86 | ||
AY437399.1 | - | 3 - 4 | 432 | 86 | ||
AY437405.1 | - | 3 - 4 | 1034 | 86 | ||
BC162118.1 | - | 3 - 4 | 930 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |