Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213151.1 | NP_998316.1 | 1 - 2 | 109 | 70 | |
GenBank mRNA | BC062383.1 | - | 1 - 2 | 109 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |