Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spint1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213152.1 | NP_998317.1 | 4 - 5 | 869 | 76 | |
XM_021467136.1 | XP_021322811.1 | 5 - 6 | 957 | 76 | ||
XM_021467137.1 | XP_021322812.1 | 5 - 6 | 958 | 76 | ||
GenBank mRNA | AY648768.1 | - | 4 - 5 | 893 | 76 | |
BC053239.1 | - | 4 - 5 | 869 | 76 | ||
BC155204.1 | - | 4 - 5 | 818 | 76 | ||
BC165390.1 | - | 3 - 4 | 746 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |