Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sec22ba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213384.1 | NP_998549.1 | 4 - 5 | 675 | 84 | |
GenBank mRNA | BC046043.1 | - | 4 - 5 | 675 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |