Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc37a4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214738.1 | NP_999903.1 | 4 - 5 | 837 | 102 | |
XM_009291487.3 | XP_009289762.1 | 5 - 6 | 1064 | 102 | ||
XM_021481373.1 | XP_021337048.1 | 5 - 6 | 1068 | 102 | ||
GenBank mRNA | BC066719.1 | - | 4 - 5 | 837 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |