Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ttc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002122.2 | NP_001002122.2 | 8 - 9 | 1208 | 134 | |
GenBank mRNA | BC071463.1 | - | 8 - 9 | 1157 | 134 | |
EH459652.1 | - | 2 - 3 | 205 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |