Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atp2b3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002472.3 | NP_001002472.2 | 6 - 7 | 1896 | 66 | |
XM_005166819.4 | XP_005166876.1 | 6 - 7 | 1814 | 66 | ||
XM_021478239.1 | XP_021333914.1 | 6 - 7 | 1814 | 66 | ||
XM_021478240.1 | XP_021333915.1 | 6 - 7 | 1816 | 66 | ||
GenBank mRNA | BC076342.1 | - | 5 - 6 | 1249 | 66 | |
EU559283.1 | - | 5 - 6 | 1245 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |