Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: phax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003995.1 | NP_001003995.1 | 2 - 3 | 684 | 93 | |
GenBank mRNA | BC080257.1 | - | 2 - 3 | 684 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |