Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: phax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003995.1 | NP_001003995.1 | 3 - 4 | 801 | 160 | |
GenBank mRNA | BC080257.1 | - | 3 - 4 | 801 | 160 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |