Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: matn3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004007.2 | NP_001004007.1 | 5 - 6 | 1100 | 65 | |
NM_001328273.1 | NP_001315202.1 | 4 - 5 | 974 | 65 | ||
GenBank mRNA | AJ629312.1 | - | 4 - 5 | 947 | 65 | |
BC080220.1 | - | 5 - 6 | 1105 | 65 | ||
BC154373.1 | - | 5 - 6 | 1100 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |