Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fn1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013261.1 | NP_001013279.1 | 21 - 22 | 3426 | 68 | |
XM_005160284.3 | XP_005160341.1 | 21 - 22 | 3461 | 68 | ||
XM_009305554.3 | XP_009303829.1 | 21 - 22 | 3462 | 68 | ||
GenBank mRNA | AY538257.1 | - | 21 - 22 | 3426 | 68 | |
AY725818.1 | - | 21 - 22 | 3441 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Danio rerio Probe spans exons |