Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hgfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013274.1 | NP_001013292.1 | 10 - 11 | 1363 | 86 | |
XM_005164742.3 | XP_005164799.1 | 10 - 11 | 2289 | 86 | ||
GenBank mRNA | BC135005.1 | - | 10 - 11 | 1366 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |