Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hgfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013274.1 | NP_001013292.1 | 11 - 12 | 1504 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |