Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spopla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013447.1 | NP_001013465.1 | 6 - 7 | 739 | 83 | |
GenBank mRNA | BC090815.1 | - | 6 - 7 | 739 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |