Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jmjd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017615.1 | NP_001017615.1 | 6 - 7 | 846 | 79 | |
XM_021467157.1 | XP_021322832.1 | 6 - 7 | 894 | 79 | ||
GenBank mRNA | BC092834.1 | - | 6 - 7 | 846 | 79 | |
BC164846.1 | - | 6 - 7 | 721 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |