Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rgs5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017821.2 | NP_001017821.1 | 3 - 4 | 419 | 85 | |
GenBank mRNA | BC092795.1 | - | 3 - 4 | 420 | 85 | |
BC165301.1 | - | 3 - 4 | 373 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |